Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, Sabinene hydrate
Supelco® Organic Reference Standard, Sabinene hydrate
Supplier:  Merck
Certificates
undefined
Supelco® Organic Reference Standard, Sabinene hydrate
Supplier:  Merck

Specifications

  • CAS No.:
    546-79-2
  • Storage conditions:
    2...30 °C
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Synonym(s):
    4-Thujanol
  • Melting point:
    58 - 62 °C
  • Molecular weight:
    154,25

Specifications

Frequently Bought Together