Supelco® Organic Reference Standard, Rhamnetin
Catalog # 17799-5MG-F
Supplier: Merck
CAS Number:
Specifications
- Pk:5 mg
- Standards:Analytical standard
- CAS No.:90-19-7
- Description:Rhamnetin
- Seq:02
- Storage conditions:2...8 °C
- Shipping conditions:STD
- MDL No.:MFCD00016931
- M-Clarity quality level:MQ100
- Synonym(s):β-Rhamnocitrin; 3,3′,4′,5-Tetrahydroxy 7-methoxyflavone; 7-Methoxyquercetin; 7-Methylquercetin; Quercetin 7-methyl ether
- Formula:C₁₆H₁₂O₇
- Molecular weight:316,26
- Storage temperature:2...8 °C
- Flash point:Not applicable
- Form:Neat
Specifications
About this item
- GHS signal: GHS07
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.