Organic reference standard, Red 2G, analytical standard, Supelco®
40462-25MG
: Merck
:
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:3734-67-6
- Description:Red 2G, ≥98,0% (HPLC)
- Formula:C₁₈H₁₃N₃Na₂O₈S₂
- Description:Red 2G
- Form:Neat
Red 2G is a compound, which belongs to the azo family of dyes, generally used in textile industry. It can mainly find applications in non-linear optical studies.
: Bottomless glass bottle. Contents are inside inserted fused cone.