Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Propionaldehyde, analytical standard, Supelco®
Organic reference standard, Propionaldehyde, analytical standard, Supelco®
Supplier:  Merck
Certificates
undefined
Organic reference standard, Propionaldehyde, analytical standard, Supelco®
Supplier:  Merck

Specifications

  • CAS No.:
    123-38-6
  • Melting point:
    –80 °C
  • Formula:
    CH₃CH₂CHO
  • Description:
    Propionaldehyde
  • Flash point:
    –40 °C
  • Boiling point:
    46…50 °C

Specifications

Frequently Bought Together