Phosphoglucose isomerase from baker's yeast (S, cerevisiae), Supelco®
Catalog # F2668-4ML
Supplier: Merck
CAS Number:
Specifications
- Pk:4 mL
- CAS No.:9001-41-6
- Description:Phosphoglucose Isomerase
- Description:Phosphoglucose Isomerase
Specifications
About this item
For use with Fructose assay Kit FA-20.