Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Paraben internal standard mix solution, analytical standard, Supelco®
Organic reference standard, Paraben internal standard mix solution, analytical standard, Supelco®
Catalog # 32125-1ML
Supplier:  Merck
CAS Number:  
Organic reference standard, Paraben internal standard mix solution, analytical standard, Supelco®
Catalog # 32125-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    50 µg/ml in acetone
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • Description:
    Paraben internal standard mix solution
  • Storage temperature:
    –20 °C
  • Description:
    Paraben internal standard mix solution
  • Flash point:
    –17 °C

Specifications

About this item