Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, L-Valine hydrochloride solution, analytical standard, Supelco®
Organic reference standard, L-Valine hydrochloride solution, analytical standard, Supelco®
Catalog # 19515-5ML-F
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, L-Valine hydrochloride solution, analytical standard, Supelco®
Catalog # 19515-5ML-F
Supplier:  Merck
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Concentration:
    100 mM amino acid in 0,1 M HCl
  • Pk:
    5 mL
  • Standards:
    Analytical standard
  • CAS No.:
    17498-50-9
  • Description:
    L-Valine hydrochloride solution
  • M-Clarity quality level:
    MQ100
  • Formula:
    C₅H₁₁NO₂ ·HCl
  • Storage temperature:
    2…8 °C
  • Description:
    L-Valine hydrochloride solution

Specifications

About this item

L-Valine hydrochloride is a semiorganic nonlinear optical material.