Organic reference standard, L-Glutamic acid monosodium salt hydrate, analytical standard, Supelco®
Catalog # 01495-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:Analytical standard
- CAS No.:142-47-2
- Description:L-Glutamic acid monosodium salt hydrate, ≥98,5% (TLC)
- Formula:C₅H₈NNaO₄·xH₂O
- Description:L-Glutamic acid monosodium salt hydrate
Specifications
About this item
Agonist at kainate, NMDA, and quisqualate receptors; an excitatory amino acid neurotransmitter.