Organic reference standard, Isoquinoline, analytical standard, Supelco®
68853-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:119-65-3
- Description:Isoquinoline, ≥97,5% (GC)
- Melting point:26...28 °C
- Formula:C₉H₇N
- Storage temperature:2...8 °C
- Description:Isoquinoline
- Boiling point:242...243 °C
- Form:Neat