Supelco® Organic Reference Standard, Hop-22(29)-ENE Solution
04626-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:1615-91-4
- Description:Hop-22(29)-ene solution
- Storage conditions:–20 °C
- Hazard class description:H225 - H304 - H315 - H336 - H410
- Shipping conditions:STD
- MDL No.:MFCD00216319
- M-Clarity quality level:MQ100
- Synonym(s):Diploptene
- Formula:C₃₀H₅₀
- Molecular weight:410,72
- Storage temperature:−20 °C
- Flash point:–12 °C - closed cup
- GHS signal: GHS02, GHS08, GHS07, GHS09
: P210 - P273 - P301 + P310 + P331 - P302 + P352