Organic reference standard, gamma-Decalactone, analytical standard, Supelco®
Catalog # 93738-1ML
Supplier: Merck
CAS Number:
Specifications
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:706-14-9
- Description:gamma-Decalactone, ≥98,0% (GC)
- Environmentally Preferable:
- Formula:C₁₀H₁₈O₂
- Description:gamma-Decalactone
- Flash point:136 °C
- Form:Neat