Organic reference standard, Ethyl acetoacetate, analytical standard, Supelco®
63954-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:141-97-9
- Description:Ethyl acetoacetate, ≥99,0% (GC)
- Melting point:−43 °C (lit.)
- Formula:C₆H₁₀O₃
- Description:Ethyl acetoacetate
- Flash point:73 °C
- Boiling point:181 °C (lit.)
- Form:Neat
Ethyl acetoacetate is derived from ethanol.
It can be used as a diluent for improving cold flow properties of biodiesel from waste cooking oil and as an additive for activating Fe₃O₄ nanoparticles for efficient catalytic oxidation of aldehydes to carboxylic acids.