Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Erythrosin B, analytical standard, Supelco®
Organic reference standard, Erythrosin B, analytical standard, Supelco®
Catalog # 87613-25MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Erythrosin B, analytical standard, Supelco®
Catalog # 87613-25MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    16423-68-0
  • Description:
    Erythrosin B, ≥98,0% (HPLC)
  • Description:
    Erythrosin B
  • Flash point:
    182 °C
  • Form:
    Neat

Specifications

About this item

Erythrosin B belongs to the class of xanthene dyes and has application as a food colorant.

It exhibits light absorption in the range of 500 - 550 nm which leads to the formation of reactive oxygen species using which it can inactivate the microorganisms, thereby acting like an anti-microbial agent.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.