Order Entry
Northern Ireland
ContactUsLinkComponent
Phytochemical standard, cryptochlorogenic acid, analytical standard, Supelco®
Phytochemical standard, cryptochlorogenic acid, analytical standard, Supelco®
  90467-5MG
 :  Merck
 :  
undefined
Phytochemical standard, cryptochlorogenic acid, analytical standard, Supelco®
  90467-5MG
 :  Merck
 :  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • Pk:
    5 mg
  • Standards:
    Analytical standard
  • CAS No.:
    905-99-7
  • Description:
    Cryptochlorogenic acid, ≥98,0% (HPLC)
  • Formula:
    C₁₆H₁₈O₉
  • Storage temperature:
    2...8 °C
  • Description:
    Cryptochlorogenic acid
  • Form:
    Neat

 

 

Cryptochlorogenic acid is a polyphenol, found in a variety of plants, which plays a crucial role in protection against various age-related diseases.

Cryptochlorogenic acid may be used as a reference standard in the determination of cryptochlorogenic acid in the leaves of Hibiscus sabdariffa using high performance liquid chromatography coupled with mass spectrometry (HPLC-MS).

 : Bottomless glass bottle. Contents are inside inserted fused cone.