Organic reference standard, Citrus Red 2, analytical standard, Supelco®
Catalog # 89774-25MG
Supplier: Merck
CAS Number:
Specifications
- Pk:25 mg
- Standards:Analytical standard
- CAS No.:6358-53-8
- Description:Citrus Red 2, ≥98,5% (HPLC)
- Formula:C₁₈H₁₆N₂O₃
- Description:Citrus Red 2
- Form:Neat
Specifications
About this item
Citrus red 2 is an artificial dye, which belongs to the mono-azo group of dyes.
- Used to color orange skins
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.