Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Citral, analytical standard, Supelco®
Organic reference standard, Citral, analytical standard, Supelco®
Catalog # 43318-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Citral, analytical standard, Supelco®
Catalog # 43318-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    5392-40-5
  • Description:
    Citral, ≥95,0% (cis + trans, GC)
  • Formula:
    C₁₀H₁₆O 
  • Storage temperature:
    2...8 °C
  • Description:
    Citral
  • Boiling point:
    229 °C(lit.)
  • Form:
    Neat

Specifications

About this item

Citral usually is a key component of lemon scented essential oils and also in volatile oils of plants like ginger, which can exist like a mixture of two isomers, cis-isomer neral and trans-isomer geranial. It can serve as potential candidate used in the treatment of tumours. It can mainly find applications as a food additive and as a fragrance in cosmetics.