Organic reference standard, Bisphenol M, analytical standard, Supelco®
Catalog # 23454-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:13595-25-0
- Description:Bisphenol M, ≥99,0% (HPLC)
- Melting point:13...139 °C (lit.)
- Formula:C₂₄H₂₆O₂
- Description:Bisphenol M
- Form:Neat