Organic reference standard, Bisphenol F, analytical standard, Supelco®
Catalog # 51453-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:620-92-8
- Description:Bisphenol F, ≥98,0% (HPLC)
- Melting point:162...164 °C (lit.)
- Formula:C₁₃H₁₂O₂
- Description:Bisphenol F
- Form:Neat