Organic reference standard, Azelaic acid, analytical standard, Supelco®
Catalog # 95054-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:123-99-9
- Description:Azelaic acid, ≥98,5% (GC)
- Environmentally Preferable:
- Melting point:106...112 °C; 109...111 °C (lit.)
- Formula:C₉H₁₆O₄
- Description:Azelaic acid
- Boiling point:286 °C/100 mmHg (lit.)
- Form:Neat
Specifications
About this item
Packaging: Bottomless glass bottle contents are inside inserted with fused cone.