Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, alpha-Zearalenol solution, analytical standard, Supelco®
Organic reference standard, alpha-Zearalenol solution, analytical standard, Supelco®
Catalog # 35406-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, alpha-Zearalenol solution, analytical standard, Supelco®
Catalog # 35406-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    10 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    36455-72-8
  • Description:
    alpha-Zearalenol solution
  • Formula:
    C₁₈H₂₄O₅
  • Storage temperature:
    –20 °C
  • Description:
    alpha-Zearalenol solution

Specifications

About this item