Organic reference standard, alpha-Terpineol, analytical standard, Supelco®
30627-100MG
: Merck
:
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:98-55-5
- Description:alpha-Terpineol, ≥90,0% (GC)
- Melting point:31...35 °C (lit.)
- Formula:C₁₀H₁₈O
- Description:alpha-Terpineol
- Boiling point:217...218 °C (lit.)
- Form:Neat
α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity of Melaleuca alternifolia (tea tree) essential oil.
The reaction rate constant of α-terpineol with the OH radical and ozone was found to be (1.9±0.5)×10⁻¹⁰ cm³ molecule⁻¹s⁻¹ and (3.0±0.2)×10⁻¹⁶ cm³ molecule⁻¹s⁻¹, respectively.