Organic reference standard, alpha-Methylbenzyl acetate, analytical standard, Supelco®
78975-1ML
: Merck
:
- Pk:1 mL
- Standards:Analytical standard
- CAS No.:93-92-5
- Description:alpha-Methylbenzyl acetate, ≥99,0% (GC)
- Formula:C₁₀H₁₂O₂
- Description:alpha-Methylbenzyl acetate
- Boiling point:94...95 °C/12 mmHg (lit.)
- Form:Neat