Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, Aflatoxin B2-13C17 solution, analytical standard, Supelco®
Organic reference standard, Aflatoxin B2-13C17 solution, analytical standard, Supelco®
Catalog # 32771-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, Aflatoxin B2-13C17 solution, analytical standard, Supelco®
Catalog # 32771-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    0.5 µg/ml in acetonitrile
  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    1217470-98-8
  • Description:
    Aflatoxin B₂-¹³C₁₇ solution
  • Formula:
    ¹³C₁₇H₁₄O₆
  • Storage temperature:
    −20 °C
  • Description:
    Aflatoxin B₂-¹³C₁₇ solution
  • Flash point:
    2 °C

Specifications

About this item

Aflatoxin B₂-¹³C₁₇ is the ¹³C isotope-labeled analog of the mycotoxin, aflatoxin.

It may be used as an analytical standard for the determination of aflatoxin B₂ in food grade gums as well as milk, milk-based infant formula, and feed samples by stable isotope dilution and liquid chromatography combined with tandem mass spectrometry (LC-MS/MS).