Organic reference standard, 7-Methoxycoumarin, analytical standard, Supelco®
42248-10MG
: Merck
:
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:531-59-9
- Description:7-Methoxycoumarin, ≥98,0% (HPLC)
- Melting point:117...121 °C (lit.)
- Formula:C₁₀H₈O₃
- Description:7-Methoxycoumarin
- Form:Neat
: Bottomless glass bottle. Contents are inside inserted fused cone.