Organic reference standard, 5-Bromo-5-nitro-1,3-dioxane, analytical standard, Supelco®
Catalog # 34391-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Standards:Analytical standard
- CAS No.:30007-47-7
- Description:5-Bromo-5-nitro-1,3-dioxane, ≥99,0% (HPLC)
- M-Clarity quality level:MQ100
- Melting point:58...60 °C
- Formula:C₄H₆BrNO₄
- Storage temperature:2…8 °C
- Description:5-Bromo-5-nitro-1,3-dioxane
- Form:Neat