Order Entry
Northern Ireland
ContactUsLinkComponent
 
Organic reference standard, 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, analytical standard, Supelco®
  44527-100MG
 :  Merck
 :  
undefined
Organic reference standard, 4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, analytical standard, Supelco®
  44527-100MG
 :  Merck
 :  

 

  • Pk:
    100 mg
  • Standards:
    Analytical standard
  • CAS No.:
    28664-35-9
  • Description:
    4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one, ≥97,0% (GC)
  • Formula:
    C₆H₈O₃
  • Description:
    4,5-Dimethyl-3-hydroxy-2,5-dihydrofuran-2-one
  • Boiling point:
    184 °C (lit.)
  • Form:
    Neat

 

 

 : Bottomless glass bottle. Contents are inside inserted fused cone.