Organic reference standard, 2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers, analytical standard, Supelco®
Catalog # 43236-100MG
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mg
- Standards:Analytical standard
- CAS No.:27043-05-6
- Description:2-Ethyl-3,5(6)-dimethylpyrazine, mixture of isomers, ≥98,0% (GC)
- Formula:C₈H₁₂N₂
- Storage temperature:2...8 °C
- Description:2-Ethyl-3,5(6)-dimethylpyrazine
- Flash point:69 °C
- Boiling point:180...181 °C (lit.)
- Form:Neat