Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, 2-(4-tert-Butylbenzyl)propionaldehyde, analytical standard, Supelco®
Organic reference standard, 2-(4-tert-Butylbenzyl)propionaldehyde, analytical standard, Supelco®
Catalog # 43884-10MG
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, 2-(4-tert-Butylbenzyl)propionaldehyde, analytical standard, Supelco®
Catalog # 43884-10MG
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    10 mg
  • Standards:
    Analytical standard
  • CAS No.:
    80-54-6
  • Description:
    2-(4-tert-Butylbenzyl)propionaldehyde, ≥96,0% (GC)
  • Formula:
    C₁₄H₂₀O
  • Storage temperature:
    2...8 °C
  • Description:
    2-(4-tert-Butylbenzyl)propionaldehyde

Specifications

About this item

2-(4-tert-Butylbenzyl)propionaldehyde is categorised under the synthetic fragrance class of compounds widely utilised in consumer products such as perfumes, after shave lotions, cosmetics and more.

2-(4-tert-Butylbenzyl)propionaldehyde can be used as a reference standard for the determination of 2-(4-tert-butylbenzyl)propionaldehyde in: Human urine samples by ultra-high performance liquid chromatography-tandem mass spectrometry (UHPLC-MS/MS) equipped with electrospray ionisation (ESI) source and multiple reaction monitoring (MRM) mode of detection, deodorants and air fresheners by sonication extraction coupled with gas chromatography-mass spectrometry (GC-MS), scented consumer products by GC-MS as well as GC in combination with olfactometry.