Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
  32401-25MG
 :  Merck
 :  
undefined
Organic reference standard, (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol, analytical standard, Supelco®
  32401-25MG
 :  Merck
 :  

 

  • Pk:
    25 mg
  • Standards:
    Analytical standard
  • CAS No.:
    342611-01-2
  • Description:
    (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol
  • Formula:
    C₃D₅H₂ClO₂
  • Description:
    (+/-)-3-Chloro-1,2-propane-1,1,2,3,3,-d5-diol
  • Form:
    Neat

 

 

3-MCPD-d5 is the deuterated form of 3-MCDP.

  • Used as an internal standard in GC-MS method for the determination of total 3-MCPD concentration from edible fats and oils.