Supelco® Organic Reference Standard, (+)-Garcinia Acid
Catalog # 44292-10MG
Supplier: Merck
CAS Number:
Specifications
- Pk:10 mg
- Standards:Analytical standard
- CAS No.:27750-13-6
- Description:(+)-Garcinia acid
- Storage conditions:2...30 °C
- Hazard class description:H319
- Shipping conditions:STD
- MDL No.:MFCD06656311
- M-Clarity quality level:MQ100
- Synonym(s):(−)-Hydroxycitric acid lactone; (2S,3S)-3-Hydroxy-5-oxotetrahydrofuran-2,3-dicarboxylic acid; Garcinia lactone
- Formula:C₆H₆O₇
- Molecular weight:190,11
- Flash point:Not applicable
Specifications
About this item
- GHS signal: GHS06
Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.
Caution: P264 - P280 - P305 + P351 + P338 - P337 + P313