Order Entry
Northern Ireland
ContactUsLinkComponent
Organic reference standard, (-)-alpha-Bisabolol, analytical standard, Supelco®
Organic reference standard, (-)-alpha-Bisabolol, analytical standard, Supelco®
Catalog # 95426-1ML
Supplier:  Merck
CAS Number:  
undefined
Organic reference standard, (-)-alpha-Bisabolol, analytical standard, Supelco®
Catalog # 95426-1ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Standards:
    Analytical standard
  • CAS No.:
    23089-26-1
  • Description:
    (-)-alpha-Bisabolol, ≥93% (GC)
  • Formula:
    C₁₅H₂₆O
  • Storage temperature:
    2…8 °C
  • Description:
    (-)-alpha-Bisabolol
  • Form:
    Neat

Specifications

About this item

(−)-α-Bisabolol is a monocyclic sesquiterpene alcohol naturally found in the essential oil of different plants like Vanillosmopsis erythropappa and Matricaria chamomilla.

It finds applications in clinical and cosmetic industries due to its pharmacological properties like anti-inflammatory, anti-irritant, antimicrobial and skin healing property. It also shows effective action against malignant tumor cells.

Packaging: Bottomless glass bottle. Contents are inside inserted fused cone.