Order Entry
Northern Ireland
ContactUsLinkComponent
DPA/Terbium set for membrane fusion assay
DPA/Terbium set for membrane fusion assay
Catalog # 80104.
Supplier:  Biotium
CAS Number:  
DPA/Terbium set for membrane fusion assay
Catalog # 80104.
Supplier:  Biotium
CAS Number:  

Specifications

  • Pk:
    1 SET
  • Seq:
    0125

Specifications

About this item

 :
Storage Temperature:Ambient
 :13798-24-8