
p-Xylene-bis(N-pyridinium bromide) (DPX)
Catalog # 80012.
Supplier: Biotium
CAS Number:
Specifications
- Pk:500 mg
- Seq:2725
Specifications
About this item
:
:422.16 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:14208-10-7
Formula:C₁₈H₁₈Br₂N₂
MDL Number:MFCD00043749