Order Entry
Northern Ireland
ContactUsLinkComponent
Pesticide Reference standard, 4,4'-DDT solution, certified reference material, Supelco®
Pesticide Reference standard, 4,4'-DDT solution, certified reference material, Supelco®
Catalog # 40124.
Supplier:  Merck
CAS Number:  
undefined
Pesticide Reference standard, 4,4'-DDT solution, certified reference material, Supelco®
Catalog # 40124.
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    5000 μg/ml in methanol
  • Pk:
    1
  • Standards:
    Certified reference material
  • CAS No.:
    50-29-3
  • Description:
    4,4′-DDT solution
  • Formula:
    C₁₄H₉Cl₅
  • Description:
    4,4′-DDT solution
  • Pk Info:
    Ampoule of 1 ml

Specifications

About this item

Certifications: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025:2005.