Supelco® Cerilliant® Organic Reference Standard, L-Ascorbic Acid (Vitamin C)-¹³C₆ solution
Catalog # V-047-1ML
Supplier: Merck
CAS Number:
Specifications
- Concentration:500 µg/ml in acetonitrile: water
- Pk:1 mL
- Standards:Certified reference material, Cerilliant®
- Description:L-Ascorbic Acid (Vitamin C)-¹³C₆ solution
- Storage conditions:–70 °C
- Hazard class description:H225 - H302 - H319
- Shipping conditions:WICE
- MDL No.:MFCD00064328
- M-Clarity quality level:MQ300
- Molecular weight:182,08
- Flash point:2 °C
- Pk Info:Ampule of 1 ml
Specifications
About this item
- GHS signal: GHS02, GHS07
Caution: P210 - P233 - P240 - P241 - P301 + P312 - P305 + P351 + P338