Fluorescein-DHPE pH-indicator, membrane stain for histology
Catalog # 60024.
Supplier: Biotium
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:1182.53 g/mol
Storage Temperature:Freezer
CAS Number:87706-98-7
Formula:C₆₄H₁₀₀N₃O₁₃PS