
Rhodamine 123 mitochondria dye, for fluorescence microscopy
Catalog # 70010.
Supplier: Biotium
CAS Number:
Specifications
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:380.82 g/mol
:GHS07
Storage Temperature:Refrigerator
CAS Number:62669-70-9
Formula:C₂₁H₁₇ClN₂O₃
MDL Number:MFCD00012664