1-Propanesulphonic acid sodium salt monohydrate ≥99.0% (by titrimetric analysis) for ion pair chromatography, Supelco®
Catalog # 81806-10G-F
Supplier: Merck
CAS Number:
Specifications
- Pk:10 g
- Seq:3236
Specifications
About this item
:
:164.16 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:304672-01-3
Formula:CH₃CH₂CH₂SO₃Na·H₂O
MDL Number:MFCD00167033