



Boron trifluoride solution ∼10% (∼1,3 M) in 1-butanol, LiChropur™ for GC derivatisation, Supelco®
Catalog # 83253-100ML-F
Supplier: Merck
CAS Number:
Specifications
- Pk:100 mL
- Seq:2440
Specifications
About this item
:
:67.81 g/mol
ADR:3(8),I
:M-Clarity™ quality level = MQ100
UN:2920
:GHS02
:GHS05
:GHS06
:GHS08
Storage Temperature:Refrigerator
CAS Number:7637-07-2
:692-39-7
Formula:BF₃
MDL Number:MFCD00011316