Ferrocenoyl chloride ≥96.0% (by titrimetric analysis), LiChropur™ derivatisation reagent, for HPLC, Supelco®
Catalog # 91032-1G
Supplier: Merck
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
:
:248.49 g/mol
Storage Temperature:Freezer
UN:3261
ADR:8,III
:GHS05
CAS Number:1293-79-4
Formula:C₁₁H₉ClFeO