Order Entry
Northern Ireland
ContactUsLinkComponent
 
Pesticide standard, (Z)-Metominostrobin, reference material, Honeywell Fluka™
Catalog # HONCR1661-10MG
CAS Number:  
Pesticide standard, (Z)-Metominostrobin, reference material, Honeywell Fluka™
Catalog # HONCR1661-10MG
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    10 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    133408-51-2
  • Description:
    (Z)-Metominostrobin
  • Formula:
    C₁₆H₁₆N₂O₃
  • Storage temperature:
    Ambient
  • Description:
    (Z)-Metominostrobin
  • Form:
    Crystalline

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery information: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.