Order Entry
Northern Ireland
ContactUsLinkComponent
Pesticide standard, Carbaryl, reference material, Honeywell Fluka™
Pesticide standard, Carbaryl, reference material, Honeywell Fluka™
Catalog # HONCR1621-100MG
CAS Number:  
Pesticide standard, Carbaryl, reference material, Honeywell Fluka™
Catalog # HONCR1621-100MG
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    100 mg
  • Standards:
    Analytical Standard
  • CAS No.:
    63-25-2
  • Description:
    Carbaryl
  • Formula:
    C₁₂H₁₁NO₂
  • Storage temperature:
    Ambient
  • Description:
    Carbaryl
  • Form:
    Crystalline

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.