Order Entry
Northern Ireland
ContactUsLinkComponent
Organic analytical standards, n-Propylacetate, reference material, Honeywell Fluka™
Organic analytical standards, n-Propylacetate, reference material, Honeywell Fluka™
Catalog # HONCR1594-5ML
CAS Number:  
Organic analytical standards, n-Propylacetate, reference material, Honeywell Fluka™
Catalog # HONCR1594-5ML
CAS Number:  

Specifications

  • Pack type:
    Glass Bottle
  • Pk:
    5 mL
  • Standards:
    Analytical Standard
  • CAS No.:
    109-60-4
  • Description:
    n-Propylacetate
  • Formula:
    CH₃COOCH₂CH₂CH₃
  • Storage temperature:
    Ambient
  • Description:
    n-Propylacetate
  • Form:
    Liquid, clear

Specifications

About this item

High quality organic reference materials produced according to ISO 9001.

Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.