Organic analytical standards, Acetonitrile, reference material, Honeywell Fluka™
Catalog # HONCR1589-5ML
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass Bottle
- Pk:5 mL
- Standards:Analytical Standard
- CAS No.:75-05-8
- Description:Acetonitrile
- Formula:CH₃CN
- Storage temperature:Ambient
- Description:Acetonitrile
- Form:Liquid
Specifications
About this item
High quality organic reference materials produced according to ISO 9001.
Delivery: Supplied with complete Certificate of Analysis (CoA) with expiry date and lot specific analytical results.