Supelco® Organic Reference Standard, Fatty Acid Methyl Esters, Saturated Straight Chains
ME10-1KT
: Merck
:
- Pk:1 KIT
- Standards:Analytical standard
- Description:Fatty acid methyl esters, saturated straight chains
- Storage conditions:RT
- Hazard class description:H226 - H410
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:43 °C
- GHS signal: GHS02, GHS05
: Kit Components: Caproic (C6:0) 1 g; Caprylic (C8:0) 1 g; Capric (C10:0) 1 g; Lauric (C12:0) 1 g; Myristic (C14:0) 1 g; Palmitic (C16:0) 1 g; Stearic (C18:0) 1 g; Arachidic (C20:0) 1 g; Behenic (C22:0) 1 g; Lignoceric (C24:0) 100 mg.
: P210 - P233 - P240 - P241 - P242 - P273