Order Entry
Northern Ireland
ContactUsLinkComponent
 
Supelco® Organic Reference Standard, F.A.M.E. MIX RAPESEED OIL, 100MG, NEAT
  O7756-1AMP
 :  Merck
 :  
Supelco® Organic Reference Standard, F.A.M.E. MIX RAPESEED OIL, 100MG, NEAT
  O7756-1AMP
 :  Merck
 :  

 

  • Pk:
    1 Ampoul
  • Standards:
    Analytical standard
  • Description:
    F.A.M.E. mix Rapeseed oil, NEAT
  • Storage conditions:
    –10...–25 °C
  • Shipping conditions:
    DICE
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    Ampule of 100 mg