Order Entry
Northern Ireland
ContactUsLinkComponent
Supelco® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Supelco® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Catalog # 46866-U
Supplier:  Merck
CAS Number:  
Supelco® Organic Reference Standard, EPA 8270 Semivolatile Internal Standard Mix
Catalog # 46866-U
Supplier:  Merck
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each component in methylene chloride
  • Pk:
    1
  • Standards:
    Certified reference material
  • Description:
    EPA 8270 Semivolatile internal standard mix
  • Storage conditions:
    2…8 °C
  • Hazard class description:
    H315 - H319 - H336 - H350 - H411
  • Shipping conditions:
    STD
  • M-Clarity quality level:
    MQ100
  • Pk Info:
    Ampule of 25 ml

Specifications

About this item

  • GHS signal: GHS08, GHS09
Caution: P202 - P261 - P273 - P302 + P352 - P305 + P351 + P338 - P308 + P313