Supelco® Organic Reference Standard, Epa 547 Glyphosate Solution
Catalog # 44690-U
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Certified reference material
- Description:EPA 547 Glyphosate Solution
- Storage conditions:2...8 °C
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Synonym(s):Glyphosate solution; N-(Phosphonomethyl)glycine
- Concentration:1000 mug/ml in H₂O