Organic reference standard, Transformer oil (PCB free), analytical standard, Supelco®
Catalog # 46956.
Supplier: Merck
CAS Number:
Specifications
- Pk:1
- Standards:Analytical standard
- Description:Transformer oil (PCB free)
- Storage temperature:RT
- Description:Transformer oil (PCB free)
- Pk Info:10×5 ml/packaging