Pesticide reference standard, Thiometon in Shellsol, PESTANAL®, analytical standard, Supelco®
Catalog # 31681-250MG
Supplier: Merck
CAS Number:
Specifications
- Pk:250 mg
- Standards:Analytical standard
- Description:Thiometon in Shellsol
- M-Clarity quality level:MQ100
- Storage temperature:2...8 °C
- Description:Thiometon in Shellsol